Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000745.3 | NP_000736.2 | 6 | 1717 | ||
NM_001307945.1 | NP_001294874.1 | 6 | 930 | |||
XM_005254142.3 | XP_005254199.1 | 6 | 1178 | |||
XM_017021881.1 | XP_016877370.1 | 6 | 1184 | |||
GenBank mRNA | AJ584705.1 | - | 1 | 285 | ||
AJ584706.1 | - | 1 | 285 | |||
BC033639.1 | - | 6 | 1679 | 5501542 | ||
M83712.1 | - | 6 | 1665 | |||
U62434.1 | - | 6 | 1671 |