Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015052.2 | NP_001015052.1 | 4 | 847 | ||
NM_001015054.2 | NP_001015054.1 | 4 | 790 | |||
NM_002434.3 | NP_002425.2 | 5 | 1040 | |||
GenBank mRNA | AL520052.3 | - | 5 | 948 | ||
AY258284.1 | - | 4 | 756 | |||
BC014991.1 | - | 4 | 786 | 4909112 | ||
L10752.1 | - | 4 | 708 | |||
M71215.1 | - | 3 | 519 | |||
M74905.1 | - | 4 | 868 | |||
M99626.1 | - | 1 | 36 | |||
S51033.1 | - | 3 | 641 | |||
X56528.1 | - | 3 | 519 |