Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PPP1CC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002710.3 | NP_002701.1 | 7 | 1578 | ||
GenBank mRNA | AK091841.1 | - | 1 | 1303 | ||
AK297882.1 | - | 7 | 1296 | |||
BC014073.2 | - | 7 | 1471 | 3956353 | ||
L07395.1 | - | 7 | 1295 | |||
X74008.1 | - | 7 | 1461 |