Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
綜合式 CRO CDMO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPP2R5C SIRNA Assays ›
Gene Symbol:
PPP2R5C
Gene Name:
protein phosphatase 2 regulatory subunit B'gamma
Gene Aliases:
B56G, PR61G
Chromosome Location:
Chr.14: 101760573 - 101927992 on Build GRCh38
UniGene ID:
Hs.368264
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
114268
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPP2R5C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001161725.1
NP_001155197.1
4
408
NM_001161726.1
NP_001155198.1
4
480
NM_002719.3
NP_002710.2
2
359
NM_178586.2
NP_848701.1
2
359
NM_178587.2
NP_848702.1
2
359
XM_005267819.1
XP_005267876.1
4
469
XM_005267820.1
XP_005267877.1
4
409
XM_005267822.3
XP_005267879.1
4
469
XM_005267823.1
XP_005267880.1
4
469
XM_005267824.1
XP_005267881.1
4
469
XM_005267826.1
XP_005267883.1
2
182
XM_005267827.1
XP_005267884.1
4
404
XM_011536914.2
XP_011535216.1
3
273
XM_011536915.2
XP_011535217.1
3
356
XM_011536916.2
XP_011535218.1
3
1355
XM_011536918.1
XP_011535220.1
3
510
XM_011536919.2
XP_011535221.1
3
492
XM_011536920.1
XP_011535222.1
3
386
XM_011536921.2
XP_011535223.1
2
275
XM_017021422.1
XP_016876911.1
3
425
XM_017021423.1
XP_016876912.1
2
359
XM_017021424.1
XP_016876913.1
3
386
GenBank mRNA
AB451341.1
-
2
219
AK131391.1
-
4
393
AK302470.1
-
4
308
AK315072.1
-
2
295
BC016183.1
-
2
257
4855950
BI560029.1
-
2
333
D26445.1
-
2
216
DA853338.1
-
2
359
L42375.1
-
2
189
U37352.1
-
2
277
Z69030.1
-
2
297
Back To Top
More Information
Panther Classification:
Molecular Function -
protein phosphatase
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
negative regulation of cell proliferation
regulation of phosphatidylinositol 3-kinase signaling
regulation of protein phosphatase type 2A activity
proteasome-mediated ubiquitin-dependent protein catabolic process
protein binding
protein phosphatase type 2A regulator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-tm9kw:80/10.239.162.147:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0