Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCF4 SIRNA Assays ›
Gene Symbol:
TCF4
Gene Name:
transcription factor 4
Gene Aliases:
E2-2, FECD3, ITF-2, ITF2, PTHS, SEF-2, SEF2, SEF2-1, SEF2-1A, SEF2-1B, SEF2-1D, TCF-4, bHLHb19
Chromosome Location:
Chr.18: 55222331 - 55635993 on Build GRCh38
UniGene ID:
Hs.605153
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
114336
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001083962.1
NP_001077431.1
9
1189
NM_001243226.2
NP_001230155.2
10
979
NM_001243227.1
NP_001230156.1
8
1023
NM_001243228.1
NP_001230157.1
9
1133
NM_001243230.1
NP_001230159.1
8
665
NM_001243231.1
NP_001230160.1
7
623
NM_001243232.1
NP_001230161.1
5
645
NM_001243233.1
NP_001230162.1
5
586
NM_001243234.1
NP_001230163.1
2
218
NM_001243235.1
NP_001230164.1
2
218
NM_001243236.1
NP_001230165.1
2
234
NM_001306207.1
NP_001293136.1
8
814
NM_001306208.1
NP_001293137.1
5
645
NM_003199.2
NP_003190.1
9
1189
XM_005266739.3
XP_005266796.2
8
829
XM_005266741.3
XP_005266798.1
9
789
XM_005266743.4
XP_005266800.1
8
650
XM_005266744.3
XP_005266801.1
8
770
XM_005266745.3
XP_005266802.1
8
632
XM_005266747.3
XP_005266804.1
7
626
XM_005266749.3
XP_005266806.1
5
417
XM_005266752.4
XP_005266809.1
5
987
XM_005266754.3
XP_005266811.1
4
411
XM_005266755.4
XP_005266812.1
5
446
XM_005266761.3
XP_005266818.1
2
730
XM_006722536.2
XP_006722599.1
9
1112
XM_006722537.2
XP_006722600.1
9
745
XM_006722538.2
XP_006722601.1
8
767
XM_011526158.2
XP_011524460.1
8
1743
XM_011526160.2
XP_011524462.1
4
796
XM_017025934.1
XP_016881423.1
8
826
XM_017025935.1
XP_016881424.1
8
829
XM_017025936.1
XP_016881425.1
8
829
XM_017025937.1
XP_016881426.1
8
583
XM_017025938.1
XP_016881427.1
9
742
XM_017025939.1
XP_016881428.1
9
792
XM_017025940.1
XP_016881429.1
9
1112
XM_017025941.1
XP_016881430.1
9
745
XM_017025942.1
XP_016881431.1
9
789
XM_017025943.1
XP_016881432.1
9
792
XM_017025944.1
XP_016881433.1
8
1047
XM_017025945.1
XP_016881434.1
8
1050
XM_017025946.1
XP_016881435.1
8
1064
XM_017025947.1
XP_016881436.1
8
650
XM_017025948.1
XP_016881437.1
8
1047
XM_017025949.1
XP_016881438.1
8
767
XM_017025950.1
XP_016881439.1
8
1047
XM_017025951.1
XP_016881440.1
7
626
XM_017025952.1
XP_016881441.1
5
640
XM_017025953.1
XP_016881442.1
5
417
XM_017025954.1
XP_016881443.1
5
1001
XM_017025955.1
XP_016881444.1
4
735
XM_017025956.1
XP_016881445.1
4
411
XM_017025957.1
XP_016881446.1
2
730
GenBank mRNA
AB209741.1
-
4
297
AK095041.1
-
5
645
AK096862.1
-
7
623
AK122765.1
-
9
1189
AK299169.1
-
8
1023
AK300038.1
-
2
164
AK300612.1
-
5
586
AK300636.1
-
4
316
AK301144.1
-
8
800
AK315074.1
-
9
1121
AK316165.1
-
4
360
AL518903.3
-
2
234
BC031056.1
-
10
940
BC125084.1
-
8
603
40117795
BC125085.1
-
8
603
40117798
DC298029.1
-
2
218
FR748208.1
-
4
232
FR748209.1
-
4
733
FR748210.1
-
8
577
FR748211.1
-
8
577
FR748212.1
-
6
397
FR748213.1
-
6
397
FR748214.1
-
7
505
FR748215.1
-
5
325
FR748216.1
-
8
571
FR748217.1
-
7
451
FR748218.1
-
3
187
FR748219.1
-
5
364
FR748220.1
-
2
97
FR748221.1
-
2
97
FR748222.1
-
2
97
M74718.1
-
2
49
M74719.1
-
9
776
X52079.1
-
6
442
Back To Top
More Information
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
DNA-templated transcription, initiation
transcription initiation from RNA polymerase II promoter
positive regulation of neuron differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
protein-DNA complex assembly
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding, RNA polymerase recruiting
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, TFIIB-class binding
TFIIB-class transcription factor binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein C-terminus binding
identical protein binding
protein homodimerization activity
bHLH transcription factor binding
protein heterodimerization activity
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-tm9kw:80/10.239.162.147:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0