Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004283.3 | NP_004274.1 | 5 | 1232 | ||
GenBank mRNA | AF019226.1 | - | 1 | 260 | ||
BC016471.1 | - | 5 | 1209 | 3861912 |