Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CREBBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004380.2 | NP_004371.2 | 5 | 1449 | ||
XM_005255124.4 | XP_005255181.1 | 5 | 2056 | |||
XM_005255125.4 | XP_005255182.1 | 5 | 2056 | |||
XM_006720848.3 | XP_006720911.1 | 5 | 2056 | |||
XM_011522381.2 | XP_011520683.1 | 4 | 938 | |||
XM_011522382.2 | XP_011520684.1 | 5 | 2056 | |||
XM_017022944.1 | XP_016878433.1 | 5 | 2056 | |||
GenBank mRNA | AK304771.1 | - | 2 | 223 | ||
U47741.1 | - | 6 | 1443 | |||
U85962.3 | - | 5 | 2063 |