Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NKX2-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166175.1 | NP_001159647.1 | 2 | 1895 | ||
NM_001166176.1 | NP_001159648.1 | 2 | 1858 | |||
NM_004387.3 | NP_004378.1 | 2 | 1603 | |||
GenBank mRNA | BC025711.1 | - | 2 | 1481 | 5225103 | |
U34962.1 | - | 2 | 1550 |