Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFKFB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145443.2 | NP_001138915.1 | 15 | 3733 | ||
NM_001282630.2 | NP_001269559.1 | 15 | 3846 | |||
NM_001314063.1 | NP_001300992.1 | 16 | 4080 | |||
NM_001323016.1 | NP_001309945.1 | 16 | 3756 | |||
NM_001323017.1 | NP_001309946.1 | 15 | 4107 | |||
NM_004566.3 | NP_004557.1 | 15 | 4057 | |||
NR_136554.1 | 12 | 3483 | ||||
XM_005252464.1 | XP_005252521.1 | 14 | 3883 | |||
XM_011519493.1 | XP_011517795.1 | 13 | 3778 | |||
XM_017016329.1 | XP_016871818.1 | 14 | 4000 | |||
GenBank mRNA | AF056320.1 | - | 17 | 3741 | ||
AK094645.1 | - | 1 | 2818 | |||
AK131307.1 | - | 12 | 3444 | |||
AL583908.1 | - | 1 | 2736 | |||
BC037258.1 | - | 15 | 3658 | |||
BC040482.1 | - | 15 | 4002 | 5271907 | ||
BC042656.1 | - | 1 | 4049 |