Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POU4F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004575.2 | NP_004566.2 | 1, 2 | 542 | ||
GenBank mRNA | AK314982.1 | - | 1, 2 | 545 | ||
BC136344.1 | - | 1, 2 | 449 | 9020331 | ||
BC136345.1 | - | 1, 2 | 449 | 9020332 | ||
BY796838.2 | - | 1, 2 | 531 | |||
DB082745.1 | - | 1, 2 | 545 | |||
EU439706.1 | - | 1, 2 | 278 | |||
U06233.1 | - | 1, 2 | 509 |