Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005513.2 | NP_005504.2 | 2 | 379 | ||
XM_011512744.2 | XP_011511046.1 | 2 | 502 | |||
XM_011512745.2 | XP_011511047.1 | 2 | 450 | |||
GenBank mRNA | AK223401.1 | - | 2 | 345 | ||
BC005917.1 | - | 2 | 328 | |||
DA498425.1 | - | 2 | 379 | |||
S67859.1 | - | 2 | 328 | |||
X63468.1 | - | 1 | 319 |