Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP3CC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243974.1 | NP_001230903.1 | 2 | 516 | ||
NM_001243975.1 | NP_001230904.1 | 2 | 516 | |||
NM_005605.4 | NP_005596.2 | 2 | 516 | |||
XM_017013610.1 | XP_016869099.1 | 2 | 309 | |||
XM_017013611.1 | XP_016869100.1 | 2 | 516 | |||
GenBank mRNA | AK055013.1 | - | 2 | 667 | ||
AK299415.1 | - | 2 | 350 | |||
AY007249.1 | - | 2 | 123 | |||
BC004864.2 | - | 2 | 403 | 3677098 | ||
S46622.1 | - | 2 | 362 |