Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190415.1 | NP_001177344.1 | 3 | 434 | ||
NM_005641.3 | NP_005632.1 | 3 | 763 | |||
NM_139315.2 | NP_647476.1 | 3 | 465 | |||
NR_033792.1 | 3 | 314 | ||||
XM_006716100.2 | XP_006716163.1 | 4 | 556 | |||
XM_006716101.2 | XP_006716164.1 | 3 | 393 | |||
XM_006716102.2 | XP_006716165.1 | 3 | 260 | |||
XM_011516542.2 | XP_011514844.1 | 4 | 535 | |||
XM_011516543.2 | XP_011514845.1 | 3 | 288 | |||
XM_017012568.1 | XP_016868057.1 | 3 | 733 | |||
GenBank mRNA | AK097781.1 | - | 4 | 373 | ||
AK300005.1 | - | 3 | 417 | |||
AK314428.1 | - | 3 | 740 | |||
AK315077.1 | - | 3 | 415 | |||
AU143362.1 | - | 4 | 554 | |||
BC018115.1 | - | 3 | 408 | 3905205 | ||
CR627390.1 | - | 3 | 292 | |||
DA150478.1 | - | 3 | 393 | |||
DA776355.1 | - | 3 | 466 | |||
DC339989.1 | - | 3 | 435 | |||
L25444.1 | - | 3 | 735 | |||
U31659.1 | - | 2 | 519 |