Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005642.2 | NP_005633.2 | 1 | 1274 | ||
GenBank mRNA | AK314837.1 | - | 1 | 1408 | ||
BC032737.1 | - | 1 | 1232 | 5539359 | ||
X97999.1 | - | 1 | 1257 |