- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: MAGEB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002363.4 | NP_002354.2 | 4 | 665 | ||
NM_177404.2 | NP_796379.1 | 2 | 508 | |||
NM_177415.2 | NP_803134.1 | 3 | 582 | |||
GenBank mRNA | AK314977.1 | - | 3 | 582 | ||
BC013772.1 | - | 2 | 456 | 3917511 | ||
BU166425.1 | - | 2 | 508 | |||
CR541762.1 | - | 1 | 345 | |||
X82539.1 | - | 4 | 668 |