Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFEB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167827.2 | NP_001161299.2 | 2 | 222 | ||
NM_001271943.1 | NP_001258872.1 | 2 | 286 | |||
NM_001271944.1 | NP_001258873.1 | 2 | 286 | |||
NM_001271945.1 | NP_001258874.1 | 2 | 197 | |||
NM_007162.2 | NP_009093.1 | 3 | 388 | |||
XM_005249411.1 | XP_005249468.1 | 2 | 321 | |||
XM_005249412.1 | XP_005249469.1 | 2 | 213 | |||
XM_006715212.3 | XP_006715275.1 | 2 | 440 | |||
XM_006715213.2 | XP_006715276.1 | 2 | 264 | |||
XM_011514915.1 | XP_011513217.1 | 2 | 443 | |||
XM_011514916.2 | XP_011513218.1 | 3 | 233 | |||
GenBank mRNA | AJ608786.1 | - | 2 | 222 | ||
AJ608787.1 | - | 2 | 269 | |||
AJ608790.1 | - | 3 | 226 | |||
AJ608791.2 | - | 2 | 374 | |||
AJ608793.1 | - | 3 | 238 | |||
AJ608794.1 | - | 2 | 402 | |||
AK095061.1 | - | 2 | 1313 | |||
AK298390.1 | - | 2 | 207 | |||
AK310063.1 | - | 2 | 444 | |||
AW294760.1 | - | 2 | 286 | |||
BC032448.1 | - | 3 | 388 | 5180066 | ||
BU527754.1 | - | 2 | 197 | |||
CT000098.1 | - | 2 | 270 | |||
M33782.1 | - | 2 | 365 |