Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018058.2 | NP_001018068.1 | 5 | 657 | ||
NM_001244583.1 | NP_001231512.1 | 4 | 353 | |||
NM_012252.3 | NP_036384.1 | 6 | 744 | |||
XM_011515963.1 | XP_011514265.1 | 8 | 931 | |||
XM_011515964.2 | XP_011514266.1 | 8 | 946 | |||
XM_011515965.2 | XP_011514267.1 | 8 | 886 | |||
XM_011515966.2 | XP_011514268.1 | 8 | 995 | |||
XM_011515969.1 | XP_011514271.1 | 7 | 874 | |||
XM_011515970.1 | XP_011514272.1 | 7 | 844 | |||
XM_011515972.2 | XP_011514274.1 | 5 | 460 | |||
XM_017011874.1 | XP_016867363.1 | 7 | 1047 | |||
XM_017011875.1 | XP_016867364.1 | 7 | 882 | |||
XM_017011876.1 | XP_016867365.1 | 4 | 319 | |||
XM_017011877.1 | XP_016867366.1 | 6 | 708 | |||
XM_017011878.1 | XP_016867367.1 | 4 | 246 | |||
GenBank mRNA | AK301479.1 | - | 8 | 936 | ||
AK313546.1 | - | 6 | 696 | |||
BC029891.1 | - | 5 | 588 | 5179088 | ||
CR933605.1 | - | 5 | 657 | |||
D43945.1 | - | 6 | 673 |