Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PPARGC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013261.3 | NP_037393.1 | 2 | 260 | ||
XM_005248131.4 | XP_005248188.1 | 2 | 1003 | |||
XM_005248132.1 | XP_005248189.1 | 2 | 182 | |||
XM_005248134.4 | XP_005248191.1 | 2 | 249 | |||
XM_011513765.2 | XP_011512067.1 | 3 | 752 | |||
XM_011513766.1 | XP_011512068.1 | 2 | 182 | |||
XM_011513769.2 | XP_011512071.1 | 2 | 249 | |||
XM_017007664.1 | XP_016863153.1 | 4 | 709 | |||
GenBank mRNA | AB061325.1 | - | 2 | 215 | ||
AF106698.1 | - | 2 | 260 | |||
AF159714.1 | - | 2 | 169 | |||
AF186379.1 | - | 2 | 179 | |||
AK296591.1 | - | 2 | 218 | |||
AK309261.1 | - | 2 | 218 | |||
JQ772116.1 | - | 3 | 548 | |||
JQ772117.1 | - | 3 | 651 | |||
JQ772118.1 | - | 4 | 716 | |||
JQ772119.1 | - | 3 | 548 | |||
JQ772120.1 | - | 3 | 548 |