Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PPARGC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013261.3 | NP_037393.1 | 13 | 3195 | ||
XM_005248131.4 | XP_005248188.1 | 13 | 3938 | |||
XM_005248132.1 | XP_005248189.1 | 13 | 3117 | |||
XM_005248134.4 | XP_005248191.1 | 11 | 2910 | |||
XM_011513765.2 | XP_011512067.1 | 14 | 3687 | |||
XM_011513766.1 | XP_011512068.1 | 14 | 3269 | |||
XM_011513767.2 | XP_011512069.1 | 14 | 5407 | |||
XM_011513768.1 | XP_011512070.1 | 13 | 3168 | |||
XM_011513770.2 | XP_011512072.1 | 12 | 3405 | |||
XM_011513771.1 | XP_011512073.1 | 12 | 3014 | |||
XM_017007664.1 | XP_016863153.1 | 15 | 3644 | |||
GenBank mRNA | AF106698.1 | - | 13 | 3183 |