Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RBPJL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281448.1 | NP_001268377.1 | 6, 7 | 702 | ||
NM_001281449.1 | NP_001268378.1 | 6, 7 | 702 | |||
NM_014276.3 | NP_055091.2 | 6, 7 | 702 | |||
XM_011528521.2 | XP_011526823.1 | 6, 7 | 709 | |||
XM_011528522.2 | XP_011526824.1 | 6, 7 | 709 | |||
XM_011528523.2 | XP_011526825.1 | 6, 7 | 709 | |||
GenBank mRNA | AB024964.1 | - | 6, 7 | 622 | ||
AB026048.1 | - | 6, 7 | 622 | |||
AB027710.1 | - | 6, 7 | 622 |