Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAZAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136264.1 | NP_001129736.1 | 2, 3 | 324 | ||
NM_001136266.1 | NP_001129738.1 | 2, 3 | 324 | |||
NM_001136269.1 | NP_001129741.1 | 2, 3 | 324 | |||
NM_014764.3 | NP_055579.1 | 2, 3 | 324 | |||
GenBank mRNA | AK095963.1 | - | 2, 3 | 183 | ||
AK290119.1 | - | 2, 3 | 224 | |||
AK293330.1 | - | 2, 3 | 222 | |||
AK303828.1 | - | 2, 3 | 224 | |||
BC002334.2 | - | 2, 3 | 206 | 3509229 | ||
BC007900.2 | - | 2, 3 | 205 | 4297784 | ||
BX647278.1 | - | 2, 3 | 222 | |||
D31767.1 | - | 2, 3 | 209 | |||
DC303613.1 | - | 2, 3 | 222 | |||
DC341917.1 | - | 2, 3 | 325 |