Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAZAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136264.1 | NP_001129736.1 | 5 | 1509 | ||
NM_001136266.1 | NP_001129738.1 | 4 | 1498 | |||
NM_001136267.1 | NP_001129739.1 | 4 | 1479 | |||
NM_001136268.1 | NP_001129740.1 | 3 | 1329 | |||
NM_014764.3 | NP_055579.1 | 4 | 1575 | |||
GenBank mRNA | AK095963.1 | - | 5 | 1368 | ||
AK125855.1 | - | 3 | 3187 | |||
AY430097.1 | - | 4 | 1474 | |||
BC002334.2 | - | 4 | 1457 | 3509229 | ||
BC007900.2 | - | 4 | 1456 | 4297784 | ||
BX647278.1 | - | 4 | 1473 | |||
D31767.1 | - | 4 | 1460 |