Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MMP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304441.1 | NP_001291370.1 | 3 | 353 | ||
NM_001304442.1 | NP_001291371.1 | 3 | 350 | |||
NM_002424.2 | NP_002415.1 | 2 | 262 | |||
XM_011542834.2 | XP_011541136.1 | 3 | 342 | |||
XM_011542835.2 | XP_011541137.1 | 4 | 429 | |||
XM_011542836.2 | XP_011541138.1 | 3 | 342 | |||
XM_017017771.1 | XP_016873260.1 | 3 | 219 | |||
GenBank mRNA | AK292659.1 | - | 2 | 262 | ||
AK303388.1 | - | 3 | 353 | |||
AK303511.1 | - | 3 | 210 | |||
AK313374.1 | - | 2 | 262 | |||
AL597614.1 | - | 2 | 262 | |||
BC074988.2 | - | 2 | 201 | 30915520 | ||
BC074989.2 | - | 2 | 201 | 30915305 | ||
BC144094.1 | - | 3 | 312 | |||
J05556.1 | - | 2 | 234 |