Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TAF7L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168474.1 | NP_001161946.1 | 5, 6 | 523 | ||
NM_024885.3 | NP_079161.3 | 5, 6 | 691 | |||
XM_005262152.1 | XP_005262209.1 | 5, 6 | 523 | |||
XM_006724664.1 | XP_006724727.1 | 5, 6 | 690 | |||
GenBank mRNA | AF285595.1 | - | 5, 6 | 525 | ||
AK026810.1 | - | 4, 5 | 669 | |||
AK308261.1 | - | 5, 6 | 693 | |||
BC043391.1 | - | 5, 6 | 677 | 6154641 | ||
DB042169.1 | - | 5, 6 | 523 |