Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ILKAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030768.2 | NP_110395.1 | 12 | 1341 | ||
XM_005246106.1 | XP_005246163.1 | 12 | 1324 | |||
XM_006712784.1 | XP_006712847.1 | 12 | 1376 | |||
XM_011511946.2 | XP_011510248.1 | 11 | 1201 | |||
XM_017005056.1 | XP_016860545.1 | 12 | 1200 | |||
XM_017005057.1 | XP_016860546.1 | 12 | 1714 | |||
XM_017005058.1 | XP_016860547.1 | 11 | 1239 | |||
GenBank mRNA | AK001043.1 | - | 12 | 1353 | ||
AK055417.1 | - | 7 | 2559 | |||
AL136850.1 | - | 12 | 1266 | |||
AY024365.1 | - | 12 | 1312 | |||
BC006576.2 | - | 12 | 1341 | 3604852 | ||
BC035746.1 | - | 11 | 1736 | |||
HQ658059.1 | - | 11 | 1234 | |||
HQ658060.1 | - | 9 | 987 |