Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CITED4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133467.2 | NP_597724.1 | 1 | 1033 | ||
GenBank mRNA | BC035496.2 | - | 1 | 1035 | 3622207 | |
BC052559.1 | - | 1 | 1033 | 6250908 |