Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300961.1 | NP_001287890.1 | 10 | 987 | ||
NM_001300962.1 | NP_001287891.1 | 9 | 912 | |||
NM_001300966.1 | NP_001287895.1 | 11 | 1079 | |||
NM_006696.3 | NP_006687.3 | 11 | 1201 | |||
NM_139199.1 | NP_631938.1 | 10 | 982 | |||
XM_005271855.2 | XP_005271912.1 | 11 | 1192 | |||
XM_005271856.2 | XP_005271913.1 | 11 | 1192 | |||
XM_005271860.3 | XP_005271917.1 | 11 | 1192 | |||
XM_005271861.3 | XP_005271918.1 | 10 | 973 | |||
XM_011543109.1 | XP_011541411.1 | 11 | 1192 | |||
XM_011543110.1 | XP_011541412.1 | 11 | 1178 | |||
XM_017008969.1 | XP_016864458.1 | 10 | 973 | |||
XM_017008970.1 | XP_016864459.1 | 11 | 1192 | |||
XM_017008971.1 | XP_016864460.1 | 11 | 1192 | |||
XM_017008973.1 | XP_016864462.1 | 10 | 973 | |||
XM_017008975.1 | XP_016864464.1 | 10 | 973 | |||
GenBank mRNA | AB209079.1 | - | 11 | 1147 | ||
AK289951.1 | - | 11 | 1161 | |||
AK293621.1 | - | 9 | 872 | |||
AK297608.1 | - | 1 | 270 | |||
AK297756.1 | - | 10 | 947 | |||
AK312333.1 | - | 11 | 1161 | |||
AL136823.1 | - | 10 | 955 | |||
BC008039.2 | - | 11 | 1143 | 3009915 | ||
BC008076.2 | - | 11 | 1143 | 3009915 | ||
X87613.1 | - | 10 | 1009 |