Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003712.3 | NP_003703.1 | 5 | 745 | ||
NM_177526.2 | NP_803545.1 | 5 | 704 | |||
NM_177543.2 | NP_808211.1 | 5 | 812 | |||
XM_011528396.2 | XP_011526698.1 | 5 | 741 | |||
GenBank mRNA | AB209696.1 | - | 2 | 5893 | ||
AF035959.1 | - | 5 | 689 | |||
AF047760.1 | - | 5 | 689 | |||
AF056083.1 | - | 5 | 689 | |||
BC002806.2 | - | 5 | 696 | 3659728 | ||
BT007021.1 | - | 5 | 646 |