Gene Symbol: PPP2R2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271899.1 | NP_001258828.1 | 3 | 379 | ||
NM_001271900.1 | NP_001258829.1 | 4 | 573 | |||
NM_001271948.1 | NP_001258877.1 | 3 | 906 | |||
NM_181674.2 | NP_858060.2 | 3 | 704 | |||
NM_181675.3 | NP_858061.2 | 3 | 426 | |||
NM_181676.2 | NP_858062.1 | 2 | 510 | |||
NM_181677.2 | NP_858063.1 | 4 | 336 | |||
NM_181678.2 | NP_858064.1 | 4 | 349 | |||
NR_073526.1 | 2 | 821 | ||||
NR_073527.1 | 3 | 458 | ||||
GenBank mRNA | AK056192.1 | - | 3 | 386 | ||
AK289717.1 | - | 3 | 355 | |||
AK294659.1 | - | 4 | 482 | |||
AK295347.1 | - | 3 | 350 | |||
AK314810.1 | - | 3 | 703 | |||
AL540315.3 | - | 1 | 39 | |||
AV726282.1 | - | 4 | 255 | |||
BC031790.1 | - | 3 | 594 | 4939981 | ||
BI669304.1 | - | 4 | 334 | |||
BX647887.1 | - | 3 | 439 | |||
DA155307.1 | - | 4 | 315 | |||
DA158596.1 | - | 3 | 467 | |||
DA365141.1 | - | 4 | 336 | |||
DA522334.1 | - | 2 | 510 | |||
DA523686.1 | - | 3 | 405 | |||
DA533318.1 | - | 3 | 426 | |||
M64930.1 | - | 2 | 632 |