Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042539.2 | NP_001036004.1 | 4 | 1378 | ||
NM_001276275.1 | NP_001263204.1 | 5 | 1193 | |||
NM_001276276.1 | NP_001263205.1 | 2 | 463 | |||
NM_002383.3 | NP_002374.2 | 4 | 1378 | |||
NR_074080.1 | 3 | 527 | ||||
XM_006721047.3 | XP_006721110.1 | 5 | 1165 | |||
GenBank mRNA | AB209061.1 | - | 1 | 1186 | ||
BC041629.1 | - | 3 | 528 | 4309842 | ||
BI198991.1 | - | 3 | 412 | |||
BX362240.2 | - | 2 | 451 | |||
D85131.1 | - | 5 | 1379 | |||
EF059746.1 | - | 4 | 1213 | |||
FJ532357.1 | - | 5 | 1183 | |||
L01420.1 | - | 3 | 625 | |||
M93339.1 | - | 4 | 1264 | |||
M94046.1 | - | 4 | 1235 | |||
U33819.1 | - | 4 | 1393 |