Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LSAMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318915.1 | NP_001305844.1 | 9 | 1521 | ||
NM_002338.4 | NP_002329.2 | 7 | 1452 | |||
XM_017006383.1 | XP_016861872.1 | 8 | 1492 | |||
XM_017006384.1 | XP_016861873.1 | 8 | 743 | |||
GenBank mRNA | AK299851.1 | - | 1336 | |||
AK315286.1 | - | 7 | 1391 | |||
BC033803.1 | - | 7 | 1445 | 5188760 | ||
U41901.1 | - | 7 | 945 |