Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003320.4 | NP_003311.2 | 4, 5 | 653 | ||
NM_177972.2 | NP_813977.1 | 3, 4 | 396 | |||
XM_005253109.3 | XP_005253166.1 | 3, 4 | 388 | |||
XM_011520344.2 | XP_011518646.1 | 3, 4 | 443 | |||
GenBank mRNA | AK293528.1 | - | 3, 4 | 324 | ||
BC075031.2 | - | 4, 5 | 445 | 30915625 | ||
BC075032.2 | - | 4, 5 | 445 | 30915418 | ||
U54644.1 | - | 3, 4 | 399 | |||
U82467.1 | - | 4, 5 | 653 |