Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292032.1 | NP_001278961.1 | 3 | 313 | ||
NM_003427.4 | NP_003418.2 | 3 | 313 | |||
XM_011514846.2 | XP_011513148.1 | 3 | 287 | |||
XM_011514847.2 | XP_011513149.1 | 3 | 425 | |||
XM_011514849.2 | XP_011513151.1 | 3 | 297 | |||
XM_011514850.2 | XP_011513152.1 | 3 | 425 | |||
XM_011514854.2 | XP_011513156.1 | 3 | 297 | |||
XM_011514855.2 | XP_011513157.1 | 3 | 425 | |||
XM_017011248.1 | XP_016866737.1 | 3 | 425 | |||
XM_017011249.1 | XP_016866738.1 | 3 | 425 | |||
XM_017011250.1 | XP_016866739.1 | 3 | 425 | |||
XM_017011251.1 | XP_016866740.1 | 3 | 425 | |||
XM_017011252.1 | XP_016866741.1 | 3 | 425 | |||
XM_017011253.1 | XP_016866742.1 | 3 | 425 | |||
XM_017011254.1 | XP_016866743.1 | 3 | 297 | |||
XM_017011255.1 | XP_016866744.1 | 3 | 425 | |||
XM_017011256.1 | XP_016866745.1 | 3 | 440 | |||
XM_017011257.1 | XP_016866746.1 | 3 | 425 | |||
GenBank mRNA | AB208784.1 | - | 3 | 590 | ||
AK295276.1 | - | 3 | 415 | |||
AK302903.1 | - | 3 | 415 | |||
AK304626.1 | - | 3 | 664 | |||
AK310877.1 | - | 3 | 385 | |||
BC000133.2 | - | 3 | 293 | 2900036 | ||
BC002549.2 | - | 3 | 294 | 3139040 | ||
BT007316.1 | - | 2 | 89 | |||
DC299240.1 | - | 3 | 313 | |||
M91592.1 | - | 3 | 252 |