Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: BTAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003972.2 | NP_003963.1 | 4 | 403 | ||
XM_011540326.1 | XP_011538628.1 | 4 | 613 | |||
XM_011540329.2 | XP_011538631.1 | 4 | 613 | |||
XM_017016878.1 | XP_016872367.1 | 3 | 291 | |||
XM_017016879.1 | XP_016872368.1 | 3 | 291 | |||
XM_017016880.1 | XP_016872369.1 | 3 | 291 | |||
GenBank mRNA | AF038362.1 | - | 4 | 422 | ||
AJ001017.2 | - | 4 | 317 | |||
AK308399.1 | - | 4 | 613 | |||
BC112201.1 | - | 4 | 387 | 8327669 |