Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LRRFIP1 SIRNA Assays ›
Gene Symbol:
LRRFIP1
Gene Name:
LRR binding FLII interacting protein 1
Gene Aliases:
FLAP-1, FLAP1, FLIIAP1, GCF-2, GCF2, HUFI-1, TRIP
Chromosome Location:
Chr.2: 237627570 - 237781647 on Build GRCh38
UniGene ID:
Hs.471779
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
115560
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LRRFIP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001137550.1
NP_001131022.1
19
1445
NM_001137551.1
NP_001131023.1
7
935
NM_001137552.1
NP_001131024.1
10
1103
NM_001137553.1
NP_001131025.1
7
935
NM_004735.3
NP_004726.2
9
1031
XM_005246141.4
XP_005246198.1
8
792
XM_005246142.2
XP_005246199.1
7
742
XM_017005253.1
XP_016860742.1
20
1507
XM_017005254.1
XP_016860743.1
20
1507
XM_017005255.1
XP_016860744.1
19
1435
XM_017005256.1
XP_016860745.1
18
1321
XM_017005257.1
XP_016860746.1
18
1372
XM_017005258.1
XP_016860747.1
18
1321
XM_017005259.1
XP_016860748.1
15
1234
XM_017005260.1
XP_016860749.1
15
1147
XM_017005261.1
XP_016860750.1
13
1084
XM_017005262.1
XP_016860751.1
13
1084
XM_017005263.1
XP_016860752.1
9
907
GenBank mRNA
AF115510.1
-
19
1314
AJ223075.1
-
9
917
AK298270.1
-
7
766
AK307485.1
-
10
833
BC001385.2
-
7
771
BC018883.2
-
7
954
BC019335.1
-
7
954
BC108913.1
-
7
760
40027210
BC108914.1
-
10
928
40027218
KP455483.1
-
7
643
U69609.1
-
7
767
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
biological_process
positive regulation of type I interferon production
negative regulation of transcription, DNA-templated
cell-cell adhesion
DNA binding
double-stranded RNA binding
protein binding
protein homodimerization activity
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline