Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005121.2 | NP_005112.2 | 2 | 364 | ||
XM_011525551.2 | XP_011523853.1 | 2 | 1065 | |||
XM_011525552.2 | XP_011523854.1 | 2 | 1065 | |||
GenBank mRNA | AF117754.1 | - | 2 | 363 | ||
BC140891.1 | - | 2 | 338 | 9021759 | ||
BP332524.1 | - | 2 | 364 | |||
BQ709424.1 | - | 1 | 120 |