Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYB SIRNA Assays ›
Gene Symbol:
MYB
Gene Name:
MYB proto-oncogene, transcription factor
Gene Aliases:
Cmyb, c-myb, c-myb_CDS, efg
Chromosome Location:
Chr.6: 135180981 - 135219173 on Build GRCh38
UniGene ID:
Hs.606320
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
115653
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MYB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130172.1
NP_001123644.1
4
444
NM_001130173.1
NP_001123645.1
4
444
NM_001161656.1
NP_001155128.1
4
444
NM_001161657.1
NP_001155129.1
4
444
NM_001161658.1
NP_001155130.1
4
444
NM_001161659.1
NP_001155131.1
4
444
NM_001161660.1
NP_001155132.1
4
444
NM_005375.3
NP_005366.2
4
444
NR_134958.1
4
444
NR_134959.1
4
444
NR_134960.1
4
444
NR_134961.1
4
444
NR_134962.1
4
444
NR_134963.1
4
444
NR_134964.1
4
444
NR_134965.1
4
444
GenBank mRNA
AF104863.1
-
4
322
AJ606317.1
-
4
444
AJ606318.1
-
4
444
AJ606319.1
-
4
444
AJ606320.1
-
4
444
AJ606321.1
-
4
444
AJ606322.1
-
4
444
AJ606323.1
-
4
444
AJ606324.1
-
4
444
AJ616235.1
-
4
359
AJ616794.1
-
1
32
AY787443.1
-
4
245
AY787444.1
-
3
173
AY787445.1
-
3
173
AY787446.1
-
4
245
AY787447.1
-
4
245
AY787448.1
-
4
245
AY787449.1
-
4
245
AY787450.1
-
4
245
AY787451.1
-
4
245
AY787452.1
-
4
245
AY787453.1
-
4
245
AY787454.1
-
4
245
AY787455.1
-
4
245
AY787456.1
-
4
245
AY787457.1
-
4
245
AY787458.1
-
4
245
AY787459.1
-
4
245
AY787460.1
-
4
245
AY787461.1
-
4
245
AY787462.1
-
4
245
AY787463.1
-
4
245
AY787464.1
-
4
245
AY787465.1
-
4
245
AY787466.1
-
4
245
AY787467.1
-
4
245
AY787468.1
-
4
245
AY787469.1
-
4
245
AY787470.1
-
4
245
AY787471.1
-
4
245
AY787472.1
-
4
245
AY787473.1
-
4
245
AY787474.1
-
4
245
AY787475.1
-
4
245
BC064955.1
-
4
396
6069320
M13665.1
-
2
107
M13666.1
-
2
107
M15024.1
-
4
358
X52125.1
-
4
442
X52126.1
-
3
248
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
mitotic cell cycle
chromatin remodeling
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of T-helper cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of histone H3-K4 methylation
positive regulation of histone H3-K9 methylation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, RNA polymerase II transcription factor recruiting
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-l9dfq:80/10.239.161.188:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0