Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308123.1 | NP_001295052.1 | 2 | 510 | ||
NM_006965.3 | NP_008896.2 | 2 | 520 | |||
XM_005258341.3 | XP_005258398.1 | 2 | 507 | |||
GenBank mRNA | AF038964.1 | - | 2 | 464 | ||
AF542097.1 | - | 1 | 341 | |||
AK291246.1 | - | 2 | 499 | |||
BC003566.1 | - | 2 | 470 | 3537160 | ||
BC016801.1 | - | 2 | 491 | 4072599 | ||
BG436261.1 | - | 2 | 501 | |||
BT006658.1 | - | 1 | 329 | |||
CR450328.1 | - | 1 | 329 | |||
U68536.1 | - | 2 | 493 |