Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012228.3 | NP_036360.3 | 3 | 372 | ||
XM_011519426.2 | XP_011517728.1 | 3 | 306 | |||
GenBank mRNA | AF122004.1 | - | 3 | 289 | ||
AF151889.1 | - | 3 | 320 | |||
BC018030.1 | - | 3 | 367 | 4799583 | ||
BC066305.1 | - | 3 | 286 | |||
BC117471.1 | - | 3 | 421 | 40126022 | ||
BC130380.1 | - | 3 | 421 | 40146411 |