Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF281 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281293.1 | NP_001268222.1 | 2 | 709 | ||
NM_001281294.1 | NP_001268223.1 | 3 | 601 | |||
NM_012482.4 | NP_036614.1 | 2 | 705 | |||
XM_017000887.1 | XP_016856376.1 | 5 | 2090 | |||
XM_017000888.1 | XP_016856377.1 | 5 | 1048 | |||
GenBank mRNA | AF125158.1 | - | 1 | 601 | ||
AJ132591.1 | - | 1 | 122 | |||
AJ132592.1 | - | 2 | 685 | |||
AK022916.1 | - | 2 | 636 | |||
AK027284.1 | - | 2 | 703 | |||
AK094701.1 | - | 3 | 536 | |||
AK297233.1 | - | 3 | 559 | |||
BC051905.1 | - | 1 | 594 | 6165292 | ||
BC060820.1 | - | 2 | 685 | 30344180 |