Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBTF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076683.1 | NP_001070151.1 | 5 | 596 | ||
NM_001076684.2 | NP_001070152.1 | 5 | 682 | |||
NM_014233.3 | NP_055048.1 | 5 | 682 | |||
NR_045058.1 | 5 | 879 | ||||
XM_006722059.3 | XP_006722122.1 | 5 | 721 | |||
XM_006722060.2 | XP_006722123.1 | 4 | 465 | |||
XM_006722061.2 | XP_006722124.1 | 5 | 549 | |||
XM_017024995.1 | XP_016880484.1 | 5 | 730 | |||
XM_017024996.1 | XP_016880485.1 | 5 | 682 | |||
XM_017024997.1 | XP_016880486.1 | 5 | 874 | |||
XM_017024998.1 | XP_016880487.1 | 5 | 590 | |||
XM_017024999.1 | XP_016880488.1 | 4 | 465 | |||
XM_017025000.1 | XP_016880489.1 | 5 | 634 | |||
XM_017025001.1 | XP_016880490.1 | 5 | 682 | |||
XM_017025002.1 | XP_016880491.1 | 5 | 562 | |||
XM_017025003.1 | XP_016880492.1 | 5 | 572 | |||
XM_017025004.1 | XP_016880493.1 | 5 | 564 | |||
GenBank mRNA | AK291733.1 | - | 5 | 507 | ||
AK292518.1 | - | 5 | 879 | |||
AK292577.1 | - | 5 | 767 | |||
AK298008.1 | - | 3 | 208 | |||
AK308321.1 | - | 4 | 659 | |||
BC031423.1 | - | 4 | 620 | |||
BC042297.1 | - | 5 | 587 | 4509695 | ||
X53390.1 | - | 4 | 533 | |||
X53461.1 | - | 4 | 533 | |||
X56687.1 | - | 5 | 565 |