Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UBTF SIRNA Assays ›
Gene Symbol:
UBTF
Gene Name:
upstream binding transcription factor, RNA polymerase I
Gene Aliases:
NOR-90, UBF, UBF-1, UBF1, UBF2
Chromosome Location:
Chr.17: 44205033 - 44221626 on Build GRCh38
UniGene ID:
Hs.89781
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
115988
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UBTF
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001076683.1
NP_001070151.1
14, 15
1718
NM_001076684.2
NP_001070152.1
14, 15
1804
NM_014233.3
NP_055048.1
15, 16
1915
NR_045058.1
15, 16
2112
XM_006722059.3
XP_006722122.1
15, 16
1954
XM_006722060.2
XP_006722123.1
14, 15
1698
XM_006722061.2
XP_006722124.1
15, 16
1782
XM_017024995.1
XP_016880484.1
15, 16
1963
XM_017024996.1
XP_016880485.1
15, 16
1915
XM_017024997.1
XP_016880486.1
15, 16
2107
XM_017024998.1
XP_016880487.1
15, 16
1823
XM_017024999.1
XP_016880488.1
14, 15
1698
XM_017025000.1
XP_016880489.1
14, 15
1756
XM_017025001.1
XP_016880490.1
14, 15
1804
XM_017025002.1
XP_016880491.1
14, 15
1684
XM_017025003.1
XP_016880492.1
14, 15
1694
XM_017025004.1
XP_016880493.1
14, 15
1686
GenBank mRNA
AK291733.1
-
15, 16
1740
AK292518.1
-
15, 16
2112
AK292577.1
-
15, 16
2000
AK296917.1
-
9, 10
1028
AK298008.1
-
12, 13
1330
AK308321.1
-
13, 14
1780
BC042297.1
-
14, 15
1709
4509695
X53390.1
-
14, 15
1766
X53461.1
-
14, 15
1766
X56687.1
-
14, 15
1687
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription from RNA polymerase I promoter
transcription from RNA polymerase I promoter
transcription initiation from RNA polymerase I promoter
transcription elongation from RNA polymerase I promoter
termination of RNA polymerase I transcription
positive regulation of transcription from RNA polymerase I promoter
regulation of glucose mediated signaling pathway
RNA polymerase I CORE element sequence-specific DNA binding
RNA polymerase I upstream control element sequence-specific DNA binding
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0