Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ST18 SIRNA Assays ›
Gene Symbol:
ST18
Gene Name:
ST18, C2H2C-type zinc finger
Gene Aliases:
NZF3, ZC2H2C3, ZC2HC10, ZNF387
Chromosome Location:
Chr.8: 52110832 - 52409879 on Build GRCh38
UniGene ID:
Hs.655499
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
116039
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ST18
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014682.2
NP_055497.1
17
2894
XM_006716487.1
XP_006716550.1
17
2661
XM_011517629.1
XP_011515931.1
18
2817
XM_011517631.1
XP_011515933.1
19
2858
XM_011517632.1
XP_011515934.1
17
2672
XM_011517633.1
XP_011515935.1
17
2665
XM_011517634.1
XP_011515936.1
17
2861
XM_011517635.1
XP_011515937.1
17
2865
XM_011517636.2
XP_011515938.1
16
2942
XM_011517637.1
XP_011515939.1
15
2399
XM_011517638.2
XP_011515940.1
14
2440
XM_011517641.1
XP_011515943.1
19
2905
XM_011517642.1
XP_011515944.1
17
2571
XM_017014047.1
XP_016869536.1
17
3021
XM_017014048.1
XP_016869537.1
17
3025
XM_017014049.1
XP_016869538.1
18
3071
XM_017014050.1
XP_016869539.1
17
2858
XM_017014051.1
XP_016869540.1
16
2865
XM_017014052.1
XP_016869541.1
18
3075
XM_017014053.1
XP_016869542.1
18
3062
XM_017014054.1
XP_016869543.1
16
3111
XM_017014055.1
XP_016869544.1
16
2702
XM_017014056.1
XP_016869545.1
19
3112
XM_017014057.1
XP_016869546.1
17
2915
XM_017014058.1
XP_016869547.1
17
2676
XM_017014059.1
XP_016869548.1
17
2862
XM_017014060.1
XP_016869549.1
18
2899
XM_017014061.1
XP_016869550.1
16
3034
XM_017014062.1
XP_016869551.1
15
2786
XM_017014063.1
XP_016869552.1
16
3332
XM_017014064.1
XP_016869553.1
16
2946
XM_017014065.1
XP_016869554.1
18
3066
XM_017014066.1
XP_016869555.1
16
2705
XM_017014067.1
XP_016869556.1
16
3115
XM_017014068.1
XP_016869557.1
17
2906
XM_017014069.1
XP_016869558.1
14
2603
XM_017014070.1
XP_016869559.1
13
2524
XM_017014071.1
XP_016869560.1
14
2772
XM_017014072.1
XP_016869561.1
18
2936
XM_017014073.1
XP_016869562.1
18
2574
XM_017014074.1
XP_016869563.1
17
2857
XM_017014075.1
XP_016869564.1
18
3108
XM_017014076.1
XP_016869565.1
18
3112
XM_017014077.1
XP_016869566.1
18
2749
XM_017014078.1
XP_016869567.1
17
3029
XM_017014079.1
XP_016869568.1
17
3420
XM_017014080.1
XP_016869569.1
15
2661
XM_017014081.1
XP_016869570.1
16
2782
XM_017014082.1
XP_016869571.1
16
2617
XM_017014083.1
XP_016869572.1
15
2457
XM_017014084.1
XP_016869573.1
13
2195
GenBank mRNA
AB011107.1
-
17
2758
BC117147.1
-
12
2151
40125698
BC117149.1
-
12
2151
40125700
BC143872.1
-
12
2146
BC143873.1
-
13
2239
Back To Top
More Information
Panther Classification:
Molecular Function -
zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-cnw2s:80/10.239.163.240:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0