Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002841.1 | NP_001002841.1 | 4 | 310 | ||
NM_002476.2 | NP_002467.1 | 3 | 257 | |||
XM_005257391.4 | XP_005257448.1 | 4 | 237 | |||
XM_011524839.2 | XP_011523141.2 | 4 | 397 | |||
XM_017024683.1 | XP_016880172.1 | 4 | 384 | |||
XM_017024684.1 | XP_016880173.1 | 3 | 401 | |||
GenBank mRNA | AF116676.2 | - | 609 | |||
BC030228.1 | - | 3 | 246 | 5226518 | ||
BU658678.1 | - | 4 | 316 | |||
M24121.1 | - | 3 | 244 | |||
M36172.1 | - | 3 | 238 | |||
X13955.1 | - | 3 | 238 |