Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF589 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016089.2 | NP_057173.2 | 4 | 531 | ||
XM_017006575.1 | XP_016862064.1 | 4 | 612 | |||
XM_017006576.1 | XP_016862065.1 | 6 | 761 | |||
XM_017006577.1 | XP_016862066.1 | 4 | 1817 | |||
XM_017006578.1 | XP_016862067.1 | 4 | 592 | |||
XM_017006579.1 | XP_016862068.1 | 6 | 717 | |||
GenBank mRNA | AB209387.1 | - | 4 | 512 | ||
AF114816.1 | - | 6 | 834 | |||
AF114817.1 | - | 6 | 834 | |||
AL832831.1 | - | 4 | 516 | |||
AY258146.1 | - | 4 | 531 | |||
BC005859.2 | - | 4 | 490 | |||
BC006247.2 | - | 4 | 511 | |||
BC048798.1 | - | 4 | 508 |