Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF571 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290314.1 | NP_001277243.1 | 5 | 455 | ||
NM_001321272.1 | NP_001308201.1 | 4 | 963 | |||
NM_016536.4 | NP_057620.3 | 4 | 355 | |||
XM_011527024.2 | XP_011525326.1 | 4 | 437 | |||
XM_017026855.1 | XP_016882344.1 | 4 | 1288 | |||
XM_017026856.1 | XP_016882345.1 | 4 | 868 | |||
XM_017026857.1 | XP_016882346.1 | 4 | 1001 | |||
XM_017026858.1 | XP_016882347.1 | 4 | 370 | |||
XM_017026859.1 | XP_016882348.1 | 4 | 807 | |||
XM_017026860.1 | XP_016882349.1 | 3 | 266 | |||
GenBank mRNA | AF161544.2 | - | 4 | 336 | ||
AK291413.1 | - | 5 | 424 | |||
BC040591.1 | - | 4 | 963 | 5269760 | ||
BC113870.1 | - | 4 | 330 | 40036719 | ||
BC114479.1 | - | 4 | 334 | 40036718 | ||
BP312315.1 | - | 4 | 355 | |||
BX537401.1 | - | 4 | 354 |