Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017797.3 | NP_060267.2 | 3 | 574 | ||
XM_005259593.1 | XP_005259650.1 | 3 | 339 | |||
GenBank mRNA | AF355797.1 | - | 3 | 574 | ||
AK225831.1 | - | 3 | 573 | |||
AY007116.1 | - | 3 | 296 | |||
BC094820.1 | - | 3 | 308 |