Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NDUFS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318051.1 | NP_001304980.1 | 2 | 145 | ||
NM_002495.3 | NP_002486.1 | 2 | 145 | |||
NR_134473.1 | 2 | 145 | ||||
NR_134474.1 | 2 | 145 | ||||
NR_134475.1 | 3 | 269 | ||||
XM_017009491.1 | XP_016864980.1 | 2 | 168 | |||
GenBank mRNA | AF020351.1 | - | 2 | 123 | ||
AK307973.1 | - | 2 | 123 | |||
BU145342.1 | - | 2 | 119 | |||
BU954271.1 | - | 2 | 137 | |||
CD692133.1 | - | 3 | 285 | |||
DA609203.1 | - | 2 | 145 | |||
DB489661.1 | - | 2 | 144 | |||
KU178226.1 | - | 2 | 115 | |||
KU178227.1 | - | 2 | 115 |