Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300823.1 | NP_001287752.1 | 7 | 1055 | ||
NM_001300824.1 | NP_001287753.1 | 7 | 1055 | |||
NM_018699.3 | NP_061169.2 | 8 | 1148 | |||
XM_005262708.3 | XP_005262765.1 | 8 | 1131 | |||
XM_011531562.2 | XP_011529864.1 | 8 | 1131 | |||
XM_011531563.2 | XP_011529865.1 | 7 | 1038 | |||
XM_011531564.2 | XP_011529866.1 | 8 | 1131 | |||
XM_011531565.2 | XP_011529867.1 | 8 | 1131 | |||
XM_011531566.2 | XP_011529868.1 | 8 | 1131 | |||
XM_011531567.2 | XP_011529869.1 | 8 | 1131 | |||
XM_011531568.2 | XP_011529870.1 | 8 | 1131 | |||
XM_011531569.2 | XP_011529871.1 | 8 | 1131 | |||
XM_011531570.2 | XP_011529872.1 | 6 | 748 | |||
XM_017007668.1 | XP_016863157.1 | 8 | 1624 | |||
XM_017007669.1 | XP_016863158.1 | 8 | 1131 | |||
XM_017007670.1 | XP_016863159.1 | 8 | 1131 | |||
XM_017007671.1 | XP_016863160.1 | 2 | 222 | |||
GenBank mRNA | AF272897.1 | - | 8 | 986 | ||
AK314293.1 | - | 7 | 1221 | |||
BC121037.1 | - | 7 | 842 | 40117856 | ||
BC121038.1 | - | 7 | 842 | 40117857 |