Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MNT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020310.2 | NP_064706.1 | 6 | 2532 | ||
XM_011523868.2 | XP_011522170.1 | 6 | 2469 | |||
XM_011523869.1 | XP_011522171.1 | 3 | 1566 | |||
XM_017024654.1 | XP_016880143.1 | 3 | 4178 | |||
GenBank mRNA | AK291596.1 | - | 7 | 2380 | ||
BC117563.1 | - | 6 | 2282 | 3904263 | ||
X96401.1 | - | 6 | 2496 |